Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This Antibody was verified by Knockout to ensure that the antibody binds to the antigen stated.
FIGURE: 1 / 2
Immunogen sequence: NPKIIRQEEG LHAGTMLTTF TAQDPDRYMQ QNIRYTKLSD PANWLKIDPV NGQITTIAVL DRESPNVKNN IYNATFLASD NGIPPMSGTG TLQIYLLDIN DNAPQVLPQE AETCETPDPN SINITALDYD IDPNAGPFAF DLPLSPVTIK RNWTITRLNG DFAQLNLKIK FLEAGIYEVP IIITDSGNPP KSNISILRVK VCQCDSNGDC TDVDRIVGAG LGTGAIIAIL LCIIILLILV LMFVVWMKRR DKERQAKQLL IDPEDDVRDN ILKYDEEGGG EEDQDYDLSQ LQQPDTVEPD A
Positive Samples: Mouse brain, Mouse heart, Rat heart; Cellular Location: Cell membrane, Single-pass type I membrane protein
N-cadherin is a 140 kDa protein belonging to a family of transmembrane molecules that mediate calcium-dependent intercellular adhesion. Cadherins are involved in controlling morphogenetic movements during development and regulate cell surface adhesion through homotypic adhesion with the same cadherin species. N-cadherin's function is dependent on its association with the actin-cytoskeleton and is mediated through interactions between the C-terminal region of N-cadherin and the cytoplasmic catenin proteins. The stability of this association is regulated by phosphorylation and dephosphorylation of beta-catenin. Further, N-cadherin is a calcium dependent cell-cell adhesion glycoprotein comprised of five extracellular cadherin repeats, a transmembrane region and a highly conserved cytoplasmic tail. The protein functions during gastrulation and is required for establishment of left-right asymmetry.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cadherin 2 type 1 N-cadherin (neuronal); cadherin 2, type 1, N-cadherin (neuronal); Cadherin-2; Cadherin2; calcium-dependent adhesion protein, neuronal; CD325; CDw325; CDw325 antigen; N-cadherin; N-cadherin 1; Neural cadherin
Gene Aliases: CD325; CDH2; CDHN; CDw325; N-cadherin; NCAD
UniProt ID: (Human) P19022, (Mouse) P15116, (Rat) Q9Z1Y3
Entrez Gene ID: (Human) 1000, (Mouse) 12558, (Rat) 83501
Molecular Function:
cadherin
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support