Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: FRKHLYLLRG LPGSGKTTLA RQLQHDFPRA LIFSTDDFFF REDGAYEFNP DFLEEAHEWN QKRARKAMRN GISPIIIDNT N
N4BP2L1 (NEDD4 Binding Protein 2 Like 1) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene includekinase activity. An important paralog of this gene isN4BP2.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: NEDD4-binding protein 2-like 1
Gene Aliases: 2410024N18Rik; AI428195; B230342M21Rik; N4bp2l1
UniProt ID: (Mouse) Q3V2Q8
Entrez Gene ID: (Mouse) 100637
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support