Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MTQQGAALQ NYNNELVKCI EELCQKREEL CRQIQEEEDE KQRLQNEVRQ LTEKLARVNE NLARKIASRN EFDRTIAETE AAYLKILESS QTLLSVLKRE AGNLTKATAP DQKSSGGRDS (1-119 aa encoded by BC000864)
SSNA1 (Sjoegren syndrome nuclear autoantigen 1), also known as N14 or NA-14 (nuclear autoantigen of 14 kDa), is a 119 amino acid nuclear and cytoplasmic protein that is widely expressed and belongs to the SSNA1 family. While most highly expressed in testis, SSNA1 is found at lower levels in peripheral blood leukocytes, spleen, colon, thymus, ovary, prostate and small intestine, and is thought to be associated with microtubule structures. SSNA1 contains an N-terminal dimeric coiled-coil domain, a C-terminal basic domain and may target spastin activity at the centrosome. The gene encoding SSNA1 maps to human chromosome 9, which houses over 900 genes and comprises nearly 4% of the human genome.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Microtubule nucleation factor SSNA1; Nuclear autoantigen of 14 kDa; Sjoegren syndrome nuclear autoantigen 1; Sjoegren syndrome nuclear autoantigen 1 homolog; Sjogren syndrome nuclear autoantigen 1; Sjogren's syndrome nuclear autoantigen 1
Gene Aliases: 1110003H09Rik; 1190004J23Rik; AA934184; MNCb-2711; N14; NA-14; NA14; SSNA1
UniProt ID: (Human) O43805, (Mouse) Q9JJ94
Entrez Gene ID: (Human) 8636, (Mouse) 68475, (Rat) 311802
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support