Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
No cross-reactivity in ELISA with Human NAALADL1.
NAALADL1 has NAALADase-like activity which is currenty unknown in function. NAALADL1 has no NAAG hydrolyzing activity, exhibits a dipeptidyl-peptidase IV type activity. In vitro, cleaves Gly-Pro-AMC.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Aminopeptidase NAALADL1; N-acetylated-alpha-linked acidic dipeptidase like protein; N-acetylated-alpha-linked acidic dipeptidase-like protein; NAALADase L; NAALADase-like 1
Gene Aliases: Gm964; Naaladasel; Naaladl1
UniProt ID: (Mouse) Q7M758
Entrez Gene ID: (Mouse) 381204
Molecular Function:
metalloprotease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support