Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Members of the BTB/POZ family of transcriptional regulators, including BTBD14B, contain a conserved motif in the N-terminal region critical for protein-protein interactions and assembly of high molecular mass complexes (Korutla et al., 2002 [PubMed 11906783]).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BEN domain containing 8; BTB (POZ) domain containing 14B; BTB/POZ domain-containing protein 14B; FLJ37383; NAC-1; nucleus accumbens associated 1, BEN and BTB (POZ) domain containing; nucleus accumbens-1; Nucleus accumbens-associated protein 1; transcriptional repressor NAC1
Gene Aliases: 2010001H03Rik; 4930511N13Rik; BEND8; BTBD14B; BTBD30; NAC-1; NAC1; NACC1
UniProt ID: (Human) Q96RE7, (Rat) O35260, (Mouse) Q7TSZ8
Entrez Gene ID: (Human) 112939, (Rat) 171454, (Mouse) 66830
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support