Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Sequence of this protein is as follows: MEMEQEKMTM NKELSPDAAA YCCSACHGDE TWSYNHPIRG RAKSRSLSAS PALGSTKEFR RTRSLHGPCP VTTFGPKACV LQNPQTIMHI QDPASQRLTW NKSPKSVLVI KKMRDASLLQ PFKELCTHLM EENMIVYVEK KVLEDPAIAS DESFGAVKKK FCTFREDYDD ISNQIDFIIC LGGDGTLLYA SSLFQGSVPP VMAFHLGSLG FLTPFSFENF QSQVTQVIEG NAAVVLRSRL KVRVVKELRG KKTAVHNGLG EKGSQAAGLD MDVGKQAMQY QVLNEVVIDR GPSSYLSNVD VYLDGHLITT VQGDGVIVST PTGSTAYAAA AGASMIHPNV PAIMITPICP HSLSFRPIVV PAGVELKIML SPEARNTAWV SFDGRKRQEI RHGDSISITT SCYPLPSICV RDPVSDWFES LAQCLHWNVR KKQAHFEEEE EEEEEG
The human trans-activation response (TAR) RNA binding protein 1 (TRBP1) was initially identified as a protein that binds to the HIV-1 TAR RNA and activates the long terminal repeat (LTR) expression in the absence and presence of the viral trans-activator Tat. This binding is enhanced by the presence of co-factors such as elongation factor 1-a (EF-1-a), polypyrimidine tract-binding protein (PTB), and the chaperonin-like protein stimulator of TAR RNA-binding proteins (SRB). TRBP1 may act to disengage RNA polymerase II from TAR during transcriptional elongation if the RNA polymerase stalls at the TAR during transcriptional elongation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ13052; NAD kinase; Poly(P)/ATP NAD kinase; ppnk
Gene Aliases: dJ283E3.1; NADK
UniProt ID: (Human) O95544
Entrez Gene ID: (Human) 65220
Molecular Function:
nucleotide kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support