Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: LLQTPPMGW LAWERFRCNI NCDEDPKNCI SEQLFMEMAD RMAQDGWRDM GYTYLNIDDC WIGGRDASGR LMPDPKRFPH GIPFLADYVH SLGLKLGIYA DMGNFTCMGY PGTTLDKVVQ DAQTFAEWKV DMLKLDGCFS TPEERAQGYP KMAAALNATG RPIAFSCSWP AYEGGLPPRV NYSLLADICN LWRNYDDIQD SWWSVLSILN WFVEHQDILQ PVAGPGHWND PDMLLIGNFG LSLEQSRAQM ALWTVLAAPL LMSTDLRTIS AQNMDILQNP LMIKINQDPL GIQGRRIHKE KSLIEVYMRP LSNKASALVF FSCRTDMPYR YHSSLGQLNF TGSVIYEAQD VYSGDIISGL RDETNFTVII NPSGVVMWYL YPIKNLEMSQ Q (22-411 aa encoded by BC000095)
NAGA (N-acetylgalactosaminidase, alpha), also known as alpha-galactosidase B or GALB, is a 411 lysosomal protein belonging to the glycosyl hydrolase 27 family that may exist as a homodimer and plays a critical role in glycolipid breakdown. NAGA encodes alpha-N-acetylgalactosaminidase, a lysosomal enzyme, which cleaves alpha-N-acetylgalactosaminyl groups from glycoconjugates. Mapping to human chromosome 22q13. 2, NAGA defects are the cause of an autosomal recessive disorder with three phenotypes, known as Schindler disease (types I, II and III) or NAGA deficiency (types I, II and III). Characterized by neurologic manifestations that range in severity, Schindler disease type I is the most severe form, followed by type III, which may have mild-to-moderate effects. Schindler disease type II, also known as Kanzaki disease, is characterized by mild intellectual impairment and angiokeratoma corporis diffusum.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Acetylgalactosaminidase, alpha-N- (alpha-galactosidase B); Alpha-galactosidase B; Alpha-N-acetylgalactosaminidase; EC 3.2.1; EC 3.2.1.49; galactosidase-alphaB; galactosidasealphaB; N-acetylgalactosaminidase, alpha-
Gene Aliases: D22S674; GALB; NAGA
UniProt ID: (Human) P17050, (Rat) Q66H12, (Mouse) Q9QWR8
Entrez Gene ID: (Human) 4668, (Rat) 315165, (Mouse) 17939
Molecular Function:
galactosidase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support