Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: MADIDNKEQS ELDQDLDDVE EVEEEETGEE TKLKARQLTV QMMQNPQILA ALQERLDGLV ETPTGYIESL PRVVKRRVNA LKNLQVKCAQ IEAKFYEEVH DLERKYAVLY
NAP1L1 is a member of the nucleosome assembly protein (NAP) family. This protein participates in DNA replication and may play a role in modulating chromatin formation and contribute to the regulation of cell proliferation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hNRP; HSP22-like protein interacting protein; NAP-1-related protein; Nucleosome assembly protein 1-like 1
Gene Aliases: NAP1; NAP1L; NAP1L1; NRP
UniProt ID: (Human) P55209
Entrez Gene ID: (Human) 4673
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support