Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: MAAQKINEGL EHLAKAEKYL KTGFLKWKPD YDSAASEYGK AAVAFKNAKQ FEQAKDACLR EAVAHENNRA LFHAAKAYEQ AGMMLKEMQK LPEAVQLIEK ASMMYLENGT PDTAAMALER AGKLIENVDP EKAVQLYQQT ANVFENEERL RQAVELLGKA SRLLVRGRRF DEAALSIQKE KNIYKEIENY PTCYKKTIAQ VLVHLHRNDY VAAERCVRES YSIPGFNGSE DCAALEQLLE GYDQQDQDQV SDVCNSPLFK YMDNDYAKLG LSLVVPGGGI KKKSPATPQA KPDGVTATAA DEEEDEYSGG LC; Positive Samples: SW480, Mouse spleen, Mouse lung, Rat brain; Cellular Location: Membrane, Peripheral membrane protein
NAPG is soluble NSF attachment protein gamma. The soluble NSF attachment proteins (SNAPs) enable N-ethyl-maleimide-sensitive fusion protein (NSF) to bind to target membranes. NSF and SNAPs appear to be general components of the intracellular membrane fusion apparatus, and their action at specific sites of fusion must be controlled by SNAP receptors particular to the membranes being fused. The product of this gene mediates platelet exocytosis and controls the membrane fusion events of this process.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: gamma SNAP; Gamma-soluble NSF attachment protein; N-ethylmaleimide sensitive fusion protein attachment protein gamma; N-ethylmaleimide-sensitive factor attachment protein gamma; N-ethylmaleimide-sensitive factor attachment protein, gamma; SNAP-gamma; soluble NSF attachment protein
Gene Aliases: 2400003O04Rik; GAMMASNAP; NAPG; SNAPG; SNARE
UniProt ID: (Human) Q99747, (Mouse) Q9CWZ7
Entrez Gene ID: (Human) 8774, (Rat) 307382, (Mouse) 108123
Molecular Function:
membrane traffic protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support