Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MALGYWRAGR ARDAAEFELF FRRCPFGGAF ALAAGLRDCV RFLRAFRLRD ADVQFLASVL PPDTDPAFFE HLRALDCSEV TVRALPEGSL AFPGVPLLQV SGPLLVVQLL ETPLLCLVSY ASLVATNAAR LRLIAGPEKR LLEMGLRRAQ GPDGGLTAST YSYLGGFDSS SNVLAGQLRG VPVAGTLAHS FVTSFSGSEV PPDPMLAPAA GEGPGVDLAA KAQVWLEQVC AHLGLGVQEP HPGERAAFVA YALAFPRAFQ GLLDTYSVWR SGLPNFLAVA LALGELGYRA VGVRLDSGDL LQQAQEIRKV FRAAAAQFQV PWLESVLIVV SNNIDEEALA RLAQEGSEVN VIGIGTSVVT CPQQPSLGGV YKLVAVGGQP RMKLTEDPEK QTLPGSKAAF RLLGSDGSPL MDMLQLAEEP VPQAGQELRV WPPGAQEPCT VRPAQVEPLL RLCLQQGQVA APPPSS
Nicotinic acid (NA; niacin) is converted by nicotinic acid phosphoribosyltransferase (NAPRT; EC 2.4.2.11) to NA mononucleotide (NaMN), which is then converted to NA adenine dinucleotide (NaAD), and finally to nicotinamide adenine dinucleotide (NAD), which serves as a coenzyme in cellular redox reactions and is an essential component of a variety of processes in cellular metabolism including response to stress (Hara et al., 2007).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FHA-HIT-interacting protein; NAPRTase; Nicotinate phosphoribosyltransferase; nicotinate phosphoribosyltransferase domain containing 1; Nicotinate phosphoribosyltransferase domain-containing protein 1; nicotinic acid phosphoribosyltransferase
Gene Aliases: FHIP; NAPRT; NAPRT1; PP3856
UniProt ID: (Human) Q6XQN6
Entrez Gene ID: (Human) 93100
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support