Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MIQQLLEVA VGPLDISPKD IVQSAIMKII SALSGGSADL GGPRDPLKVL EGVVAAVHTS VDKGEELVSP EDLLEWLRPF CADDAWPVRP RIHVLQILGQ SFHLTEEDSK LLVFFRTEAI LKASWPQRQV DIADIENEEN RYCLFMELLE SSHHEAEFQH LVLLLQAWPP MKSEYVITNN PWVRLATVML TRCTMENKEG LGNEVLKMCR SLYNTKQMLP AEGVKELCLL LLNQSLLLPS LKLLLESRDE HLHEMALEQI TAVTTVNDSN CDQELLSLLL DAKLLVKCVS TPFYPRIVDH LLASLQQGRW DAEELGRHLR EAGHEAEAGS LLLAVRGTHQ AFRTFSTALR AAQHWV (2017-2371 aa encoded by BC051792)
Negatively regulates nitric oxide production by inducing NOS1 and NOS3 translocation to actin cytoskeleton and inhibiting their enzymatic activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: NAG/BC035112 fusion; NAG/FAM49A fusion; NBAS subunit of NRZ tethering complex; neuroblastoma amplified sequence; Neuroblastoma-amplified gene protein; neuroblastoma-amplified protein; neuroblastoma-amplified protein-like; Neuroblastoma-amplified sequence
Gene Aliases: 4933425L03Rik; ILFS2; NAG; NBAS; SOPH
UniProt ID: (Human) A2RRP1
Entrez Gene ID: (Human) 51594, (Rat) 690073, (Mouse) 71169
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support