Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Sequence of this protein is as follows: MLRVLVGAVL PAMLLAAPPP INKLALFPDK SAWCEAKNIT QIVGHSGCEA KSIQNRACLG QCFSYSVPNT FPQSTESLVH CDSCMPAQSM WEIVTLECPG HEEVPRVDKL VEKILHCSCQ ACGKEPSHEG LSVYVQGEDG PGSQPGTHPH PHPHPHPGGQ TPEPEDPPGA PHTEEEGAED
This gene product is the founding member of the evolutionarily conserved CAN family of proteins, which contain a domain resembling the CTCK motif found in a number of signaling molecules. These proteins are secreted, and act as BMP antagonists by binding to BMPs and preventing them from interacting with their receptors. They may thus play an important role during growth and development. Alternatively spliced transcript variants encoding distinct isoforms have been identified for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DAN domain family member 1; differential screening-selected gene aberrant in neuroblastoma; N03; neuroblastoma candidate region, suppression of tumorigenicity 1; Neuroblastoma suppressor of tumorigenicity 1; Protein N03; Zinc finger protein DAN
Gene Aliases: D1S1733E; D4H1S1733E; DAN; Dana; DAND1; NB; NBL1; NO3
UniProt ID: (Human) P41271, (Mouse) Q61477
Entrez Gene ID: (Human) 4681, (Mouse) 17965
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support