Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Sequence of this protein is as follows: MEPQVTLNVT FKNEIQSFLV SDPENTTWAD IEAMVKVSFD LNTIQIKYLD EENEEVSINS QGEYEEALKM AVKQGNQLQM QVHEGHHVVD EAPPPVVGAK RLAARAGKKP LAHYSSLVRV LGSDMKTPED PAVQSFPLVP CDTDQPQDKP PDWFTSYLET FREQVVNETV EKLEQKLHEK LVLQNPSLGS CPSEVSMPTS EETLFLPENQ FSWHIACNNC QRRIVGVRYQ CSLCPSYNIC EDCEAGPYGH DTNHVLLKLR RPVVGSSEPF CHSKYSTPRL PAALEQVRLQ KQVDKNFLKA EKQRLRAEKK QRKAEVKELK KQLKLHRKIH LWNSIHGLQS PKSPLGRPES LLQSNTLMLP LQPCTSVMPM LSAAFVDENL PDGTHLQPGT KFIKHWRMKN TGNVKWSADT KLKFMWGNLT LASTEKKDVL VPCLKAGHVG VVSVEFIAPA LEGTYTSHWR LSHKGQQFGP RVWCSIIVDP FPSEESPDNI EKGMISSSKT DDLTCQQEET FLLAKEERQL GEVTEQTEGT AACIPQKAKN VASERELYIP SVDLLTAQDL LSFELLDINI VQELERVPHN TPVDVTPCMS PLPHDSPLIE KPGLGQIEEE NEGAGFKALP DSMVSVKRKA ENIASVEEAE EDLSGTQFVC ETVIRSLTLD AAPDHNPPCR QKSLQMTFAL PEGPLGNEKE EIIHIAEEEA VMEEEEDEED EEEEDELKDE VQSQSSASSE DYIIILPECF DTSRPLGDSM YSSALSQPGL ERGAEGKPGV EAGQEPAEAG ERLPGGENQP QEHSISDILT TSQTLETVPL IPEVVELPPS LPRSSPCVHH HGSPGVDLPV TIPEVSSVPD QIRGEPRGSS GLVNSRQKSY DHSRHHHGSS IAGGLVKGAL SVAASAYKAL FAGPPVTAQP IISEDQTAAL MAHLFEMGFC DRQLNLRLLK KHNYNILQVV TELLQLNNND WYSQRY
The protein encoded by this gene was originally identified as an ovarian tumor antigen monitored in ovarian cancer. The encoded protein contains a B-box/coiled coil motif, which is present in many genes with transformation potential, but the function of this protein is unknown. This gene is located on a region of chromosome 17q21.1 that is in close proximity to tumor suppressor gene BRCA1. Three alternatively spliced variants encoding the same protein have been identified for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: B-box protein; Cell migration-inducing gene 19 protein; Membrane component chromosome 17 surface marker 2; membrane component, chromosome 17, surface marker 2 (ovarian carcinoma antigen CA125); migration-inducing protein 19; neighbor of BRCA1 gene 1; Neighbor of BRCA1 gene 1 protein; Next to BRCA1 gene 1 protein; Protein 1A1-3B
Gene Aliases: 1A1-3B; 1A13B; IAI3B; KIAA0049; M17S2; MIG19; NBR1
UniProt ID: (Human) Q14596
Entrez Gene ID: (Human) 4077
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support