Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MSLLLSFYLL GLLVSSGQAL LQVTISLSKV ELSVGESKFF TCTAIGEPES IDWYNPQGEK IISTQRVVVQ KEGVRSRLTI YNANIEDAGI YRCQATDAKG QTQEATVVLE IYQKLTFREV VSPQEFKQGE DAEVVCRVSS SPAPAVSWLY HNEEVTTISD NRFAMLANNN LQILNINKSD EGIYRCEGRV EARGEIDFRD IIVIVNVPPA ISMPQKSFNA TAERGEEMTF SCRASGSPEP AISWFRNGKL IEENEKYILK GSNTELTVRN IINSDGGPYV CRATNKAGED EKQAFLQVFV QPHIIQLKNE TTYENGQVTL VCDAEGEPIP EITWKRAVDG FTFTEGDKSL DGRIEVKGQH GSSSLHIKDV KLSDSGRYDC EAASRIGGHQ KSMYLDIEYA PKFISNQTIY YSWEGNPINI SCDVKSNPPA SIHWRRDKLV LPAKNTTNLK TYSTGRKMIL EIAPTSDNDF GRYNCTATNH IGTRFQEYIL ALADVPSSPY GVKIIELSQT TAKVSFNKPD SHGGVPIHHY QVDVKEVASE IWKIVRSHGV QTMVVLNNLE PNTTYEIRVA AVNGKGQGDY SKIEIFQTLP VREPSPPSIH GQPSSGKSFK LSITKQDDGG APILEYIVKY RSKDKEDQWL EKKVQGNKDH IILEHLQWTM GYEVQITAAN RLGYSEPTVY EFSMPPKPNI IKDTLFNGLG LGAVIGLGVA ALLLILVVTD VSCFFIRQCG LLMCITRRMC GKKSGSSGKS KELEEGKAAY LKDGSKEPIV EMRTEDERVT NHEDGSPVNE PNETTPLTEP EKLPLKEEDG KEALNPETIE IKVSNDIIQS KEDDSKA
NCAM2 (neural cell adhesion molecule 2) is a cell adhesion molecule belonging to the immunoglobulin superfamily. It is differentially expressed in the developing and mature olfactory epithelium and is involved in the guidance of developing olfactory sensory neurons to the glomeruli of the olfactory bulb.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: N-CAM-2; NCAM-2; Neural cell adhesion molecule 2
Gene Aliases: NCAM2; NCAM21
UniProt ID: (Human) O15394
Entrez Gene ID: (Human) 4685
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support