Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAPQMYEFH LPLSPEELLK SGGVNQYVVQ EVLSIKHLPP QLRAFQAAFR AQGPLAMLQH FDTIYSILHH FRSIDPGLKE DTLEFLIKVV SRHSQELPAI LDDTTLSGSD RNAHLNALKM NCYALIRLLE SFETMASQTN LVDLDLGGKG KKARTKAAHG FDWEEERQPI LQLLTQLLQL DIRHLWNHSI IEEEFVSLVT GCCYRLLENP TINHQKNRPT REAITHLLGV ALTRYNHMLS ATVKIIQMLQ HFEHLAPVLV AAVSLWATDY GMKSIVGEIV REIGQKCPQE LSRDPSGTKG FAAFLTELAE RVPAILMSSM CILLDHLDGE NYMMRNAVLA AMAEMVLQVL SG (1-351 aa encoded by BC028182)
Regulatory subunit of the condensin complex, a complex required for conversion of interphase chromatin into mitotic-like condense chromosomes. The condensin complex probably introduces positive supercoils into relaxed DNA in the presence of type I topoisomerases and converts nicked DNA into positive knotted forms in the presence of type II topoisomerases. May target the condensin complex to DNA via its C-terminal domain (PubMed:11136719). May promote the resolution of double-strand DNA catenanes (intertwines) between sister chromatids. Condensin-mediated compaction likely increases tension in catenated sister chromatids, providing directionality for type II topoisomerase-mediated strand exchanges toward chromatid decatenation. Required for decatenation of non-centromeric ultrafine DNA bridges during anaphase. Early in neurogenesis, may play an essential role to ensure accurate mitotic chromosome condensation in neuron stem cells, ultimately affecting neuron pool and cortex size (PubMed:27737959). [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: chromosome condensation related SMC associated protein 1; Chromosome condensation-related SMC-associated protein 1; Chromosome-associated protein D2; Condensin complex subunit 1; hCAP-D2; mCAP-D2; Non-SMC condensin I complex subunit D2; non-SMC condensin I complex, subunit D2; XCAP-D2 homolog
Gene Aliases: 2810406C15Rik; 2810465G24Rik; CAP-D2; CAPD2; CNAP1; hCAP-D2; KIAA0159; mKIAA0159; NCAPD2; RGD1562596
UniProt ID: (Human) Q15021, (Mouse) Q8K2Z4
Entrez Gene ID: (Human) 9918, (Rat) 362438, (Mouse) 68298
Molecular Function:
chromatin/chromatin-binding, or -regulatory protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support