Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: TTFQNEDEKN KEVYMTPLRG VKATQASKST QLKTNRGQRK VTVSARTNRR CQTAEADSES DHEVPEPESE MKMRLPRRAK TAALEKSKLN LAQFLNEDLS
This gene encodes a subunit of the condensin complex, which is responsible for the condensation and stabilization of chromosomes during mitosis and meiosis. Phosphorylation of the encoded protein activates the condensin complex. There are pseudogenes for this gene on chromosomes 8 and 15. Alternative splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: chromosome condensation protein G; Chromosome-associated protein G; Condensin complex subunit 3; Condensin subunit CAP-G; FLJ12450; hCAP-G; Melanoma antigen NY-MEL-3; MGC126525; Non-SMC condensin I complex subunit G; non-SMC condensin I complex, subunit G; XCAP-G homolog
Gene Aliases: CAPG; CHCG; NCAPG; NY-MEL-3; NYMEL3; YCG1
UniProt ID: (Human) Q9BPX3
Entrez Gene ID: (Human) 64151
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support