Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: SRRRHSDEND GGQPHKRRKT SDANETEDHL ESLICKVGEK SACSLESNLE GLAGVLEADL PNYKSKILRL LCTVARLLPE KLTIYTTLVG LLNARNYN
May be involved in protein trafficking from the plasma membrane to the early endosome (EE) as well as in homotypic fusion of endocytic organelles. Mediates the endocytic trafficking from early endosomes to late endosomes and lysosomes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 80 kDa nuclear cap-binding protein; CBP80; MGC2087; NCBP 80 kDa subunit; nuclear cap binding protein subunit 1, 80kD; nuclear cap binding protein subunit 1, 80kDa; Nuclear cap-binding protein subunit 1; RP11-546O6.1
Gene Aliases: CBP80; NCBP; NCBP1; Sto1
UniProt ID: (Human) B2R718
Entrez Gene ID: (Human) 4686
Molecular Function:
RNA metabolism protein
RNA processing factor
RNA splicing factor
nucleic acid metabolism protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support