Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: IAKWDYTAQ QDQELDIKKN ERLWLLDDSK TWWRVRNAAN RTGYVPSNYV ERKNSLKKGS LVKNLKDTLG LGKTRRKTSA RDASPTPSTD AEYPANGSGA DRIYDLNIPA FVKFAYVAER EDELSLVKGS RVTVMEKCSD GWWRGSYNGQ IGWFPSNYVL EEVDEAAAES PSFLSLRKGA SLSNGQGSRV LHVVQTLYPF SSVTEEELNF EKGETMEVIE KPENDPEWW (8-235 aa encoded by BC000103 )
The NCK family of SH2/SH3 adaptor proteins consists of two members, NCK1 and NCK2, which couple tyrosine kinase signaling, including the EGF and PDGF receptor-pathways, to downstream signaling proteins. The NCK2 protein binds human PDGFR-b (Tyr-1009); overexpression of NCK2 inhibits PDGF-induced membrane ruffling and lamellipod formation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cytoplasmic protein NCK2; Growth factor receptor-bound protein 4; NCK adaptor protein 2; Nck-2; non-catalytic region of tyrosine kinase adaptor protein 2; noncatalytic region of tyrosine kinase, beta; SH2/SH3 adaptor protein NCK-beta
Gene Aliases: 4833426I10Rik; GRB4; Nck-2; NCK2; NCKbeta
UniProt ID: (Human) O43639, (Mouse) O55033
Entrez Gene ID: (Human) 8440, (Mouse) 17974, (Rat) 316369
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support