Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MGKSNSKLK PEVVEELTRK TYFTEKEVQQ WYKGFIKDCP SGQLDAAGFQ KIYKQFFPFG DPTKFATFVF NVFDENKDGR IEFSEFIQAL SVTSRGTLDE KLRWAFKLYD LDNDGYITRN EMLDIVDAIY QMVGNTVELP EEENTPEKRV DRIFAMMDKN ADGKLTLQEF QEGSKADPSI VQALSLYDGL V (1-190 aa encoded by BC004856)
This gene is a member of the neuronal calcium sensor gene family, which encode calcium-binding proteins expressed predominantly in neurons. The protein encoded by this gene regulates G protein-coupled receptor phosphorylation in a calcium-dependent manner and can substitute for calmodulin. The protein is associated with secretory granules and modulates synaptic transmission and synaptic plasticity. Multiple transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: frequenin; Frequenin homolog; Frequenin-like protein; Frequenin-like ubiquitous protein; NCS-1; Neuronal calcium sensor 1; neuronal calcium sensor-1
Gene Aliases: 9430075O15Rik; A730032G13Rik; AI836659; FLUP; FREQ; Mfreq; NCS-1; NCS1
UniProt ID: (Human) P62166, (Mouse) Q8BNY6, (Rat) P62168
Entrez Gene ID: (Human) 23413, (Mouse) 14299, (Rat) 65153
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support