Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 5
The recommended shelf life for this product is 1 year from date of receipt.
This gene encodes a protein that mediates ligand-independent transcription repression of thyroid-hormone and retinoic-acid receptors by promoting chromatin condensation and preventing access of the transcription machinery. It is part of a complex which also includes histone deacetylases and transcriptional regulators similar to the yeast protein Sin3p. This gene is located between the Charcot-Marie-Tooth and Smith-Magenis syndrome critical regions on chromosome 17. Alternate splicing results in multiple transcript variants. Pseudogenes of this gene are found on chromosomes 17 and 20.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: h KIAA1047; hCIT529I10; MGC104216; N-CoR; N-CoR1; nuclear receptor corepressor; Nuclear receptor corepressor 1; protein phosphatase 1, regulatory subunit 109; retinoid X receptor interacting protein 13; retinoid X receptor-interacting protein 13; thyroid hormone- and retinoic acid receptor-associated corepressor 1
Gene Aliases: 5730405M06Rik; A230020K14Rik; hN-CoR; KIAA1047; mKIAA1047; N-CoR; N-CoR1; NCOR1; PPP1R109; RIP13; Rxrip13; TRAC1
UniProt ID: (Human) O75376
Entrez Gene ID: (Human) 9611, (Mouse) 20185
Molecular Function:
chromatin/chromatin-binding, or -regulatory protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support