Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: WKKHSTFGDV RKLITEEFVQ MNYLKYQRVP YVEPPEYEFF WGSRASREIT KMQIMEFLAR VFKKDPQAWP SRYREALEEA RALREANPTA HYPRSSVSED
This intronless gene is located in the Prader-Willi syndrome deletion region. It is an imprinted gene and is expressed exclusively from the paternal allele.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Necdin; necdin homolog; necdin, melanoma antigen (MAGE) family member; necdin-like protein; Prader-Willi syndrome chromosome region
Gene Aliases: HsT16328; NDN; PWCR
UniProt ID: (Human) Q99608
Entrez Gene ID: (Human) 4692
Molecular Function:
scaffold/adaptor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support