Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: MPECWDGEHD IETPYGLLHV VIRGSPKGNR PAILTYHDVG LNHKLCFNTF FNFEDMQEIT KHFVVCHVDA PGQQVGASQF PQGYQFPSME QLAAMLPSVV QHFGFKYVIG IGVGAGAYVL AKFALIFPDL VEGLVLVNID PNGKGWIDWA ATKLSGLTST LPDTVLSHLF SQEELVNNTE LVQSYRQQIG NVVNQANLQL FWNMYNSRRD LDINRPGTVP NAKTLRCPVM LVVGDNAPAE DGVVECNSKL DPTTTTFLKM ADSGGLPQVT QPGKLTEAFK YFLQGMGYMP SASMTRLARS RTASLTSASS VDGSRPQACT HSESSEGLGQ VNHTMEVSC
This gene is a member of the N-myc downregulated gene family which belongs to the alpha/beta hydrolase superfamily. The protein encoded by this gene is a cytoplasmic protein that may be involved in the regulation of mitogenic signaling in vascular smooth muscles cells. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Brain development-related molecule 1; DKFZp686I1615; FLJ30586; FLJ42011; MGC19632; N-myc downstream regulated 1, related sequence; N-myc downstream-regulated gene 4 protein; Protein Ndr4; Protein NDRG4; SMAP-8; smooth muscle-associated protein 8; Vascular smooth muscle cell-associated protein 8
Gene Aliases: BDM1; D8Bwg1337e; KIAA1180; Ndr1-rs; Ndr4; NDRG4; R74996; SMAP-8; SMAP8
UniProt ID: (Human) Q9ULP0, (Mouse) Q8BTG7
Entrez Gene ID: (Human) 65009, (Mouse) 234593
Molecular Function:
serine protease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support