Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAAAAASRGV GAKLGLREIR IHLCQRSPGS QGVRDFIEKR YVELKKANPD LPILIRECSD VQPKLWARYA FGQETNVPLN NFSADQVTRA LENVL; Positive Samples: Mouse kidney, Mouse heart; Cellular Location: Matrix side, Mitochondrion inner membrane, Peripheral membrane protein
The NDUFA2 gene encodes one of the accessory subunits of complex I, the first and largest complex of the mitochondrial respiratory chain.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CCDC76; CI-B8; complex I B8 subunit; Complex I-B8; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 2, 8kDa; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 2; NADH-ubiquinone oxidoreductase B8 subunit; NADH-ubiquinone oxidoreductase subunit CI-B8
Gene Aliases: AV000592; B8; C1-B8; CD14; CI-B8; CIB8; NDUFA2
UniProt ID: (Human) O43678, (Mouse) Q9CQ75
Entrez Gene ID: (Human) 4695, (Rat) 291660, (Mouse) 17991
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support