Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAGASLGAR FYRQIKRHPG IIPMIGLICL GMGSAALYLL RLALRSPDVC WDRKNNPEPW NRLSPNDQYK FLAVSTDYKK LKKDRPDF (1-87 aa encoded by BC011910)
NDUFA4L2 is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 4-like 2; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 4-like 2; NADH-ubiquinone oxidoreductase MLRQ subunit homolog; NADH:ubiquinone oxidoreductase MLRQ subunit homolog; NUOMS
Gene Aliases: NDUFA4L2; NUOMS
UniProt ID: (Human) Q9NRX3
Entrez Gene ID: (Human) 56901
Molecular Function:
oxidoreductase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support