Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
NDUFA9 (NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 9, mitochondrial) is also named as NDUFS2L, CI-39kD and belongs to the complex I NDUFA9 subunit family. It is involved in stabilizing the junction between membrane and matrix arms of complex I, a late assembly step critical for complex I biogenesis and activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CI-39kD; complex I 39kDa subunit; Complex I-39kD; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 9, 39kDa; NADH dehydrogenase (ubiquinone) Fe-S protein 2-like (NADH-coenzyme Q reductase); NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 9, mitochondrial; NADH-ubiquinone oxidoreductase 39 kDa subunit; ndufa9 {ECO:0000312|EMBL:AAH91192.1, ECO:0000312|RGD:1307307}; short chain dehydrogenase/reductase family 22E, member 1; Sperm flagella protein 3
Gene Aliases: 1010001N11Rik; CC6; CI-39k; CI39k; NDUFA9; NDUFS2L; SDR22E1
UniProt ID: (Human) Q16795, (Rat) Q5BK63, (Mouse) Q99JP9
Entrez Gene ID: (Human) 4704, (Rat) 362440, (Mouse) 66108
Molecular Function:
oxidoreductase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support