Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: IKSIPKGKIS IVEALTLLNN HKLFPETWTA EKIMQEYQLE QKDVNSLLKY FVTFEVEIFP PEDKKAIRSK
Antigen sequence identity: Mouse 83%; Rat 83%
NADH:ubiquinone oxidoreductase (complex I) catalyzes the transfer of electrons from NADH to ubiquinone (coenzyme Q) in the first step of the mitochondrial respiratory chain, resulting in the translocation of protons across the inner mitochondrial membrane. This gene encodes a complex I assembly factor. Mutations in this gene are a cause of mitochondrial complex I deficiency.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Hormone-regulated proliferation-associated protein of 20 kDa; hormone-regulated proliferation-associated protein, 20 kDa; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, assembly factor 4; NADH dehydrogenase (ubiquinone) complex I, assembly factor 4; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex assembly factor 4
Gene Aliases: bA22L21.1; C6orf66; HRPAP20; HSPC125; My013; NDUFAF4
UniProt ID: (Human) Q9P032
Entrez Gene ID: (Human) 29078
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support