Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Immunogen sequence: ESAGADTRPT VRPRNDVAHK QLSAFGEYVA EILPKYVQQV QVSCFNELEV CIHPDGVIPV LTFLRDHTNA QFKSLVDLTA VDVPTRQNRF EIVYNLLSLR FNSRIRVKTY TDELTPIESA VSVFKAANWY EREIWDMFGV FFANHPDLRR ILTDYGFEGH PFRKDFPLSG YVELRYDDEV KRVVAEPVEL AQEFRKFDLN SPWEAFPVYR QPPESLKLEA GDKKPDAK
This gene encodes one of the iron-sulfur protein components of mitochondrial NADH:ubiquinone oxidoreductase. Mutations in this gene are associated with Leigh syndrome resulting from mitochondrial complex I deficiency.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CI-30kD; complex I 30kDa subunit; Complex I-30kD; NADH dehydrogenase (ubiquinone) Fe-S protein 3, 30kDa (NADH-coenzyme Q reductase); NADH dehydrogenase [ubiquinone] iron-sulfur protein 3, mitochondrial; NADH dehydrogenase-ubiquinone 30 kDa subunit; NADH-ubiquinone oxidoreductase 30 kDa subunit
Gene Aliases: 0610010M09Rik; CI-30; CI-30kD; NDUFS3
UniProt ID: (Human) O75489, (Mouse) Q9DCT2
Entrez Gene ID: (Human) 4722, (Mouse) 68349, (Rat) 295923
Molecular Function:
dehydrogenase
oxidoreductase
metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support