Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Immunogen sequence: TYKYVNMQDP EMDMKSVTDR AARTLLWTEL FRGLGMTLSY LFREPATINY PFEKGPLSPR FRGEHALRRY PSGEERCIAC KLCEAICPAQ AITIEAEPRA DGSRRTTRYD IDMTKCIYCG FCQEACPVDA IVEGPNFEFS TETHEELLYN KEKLLNNGDK WEAEIAANIQ ADYLYR
Ndufs8 gene ontology annotations related to this gene include 4 iron, 4 sulfur cluster binding; oxidoreductase activity, acting on NAD(P)H.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CI-23kD; Complex I-23kD; NADH dehydrogenase [ubiquinone] iron-sulfur protein 8, mitochondrial; NADH-ubiquinone oxidoreductase 23 kDa subunit
Gene Aliases: BC021616; CI-23kD; Ndufs8; TYKY
UniProt ID: (Mouse) Q8K3J1
Entrez Gene ID: (Mouse) 225887
Molecular Function:
oxidoreductase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support