Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: AGVASSLSEK IADSIGNNRQ NAPLTSIQIR FIQNMIQETL DDFREACHRD IVNLQVEMIK QFHMQLNEMH SLLERYSVNE GLVAEIERLR EENKRLRAHF
Neural precursor cell expressed developmentally down-regulated protein 1 (NEDD1) is required for mitosis progression. NEDD1 promotes the nucleation of microtubules from the spindle.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: NEDD-1; Neural precursor cell expressed developmentally down-regulated protein 1; Protein NEDD1
Gene Aliases: GCP-WD; Nedd-1; NEDD1; TUBGCP7
UniProt ID: (Human) Q8NHV4, (Mouse) P33215
Entrez Gene ID: (Human) 121441, (Mouse) 17997
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support