Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Recommended positive controls: 293T, A431, H1299, HeLa, Molt-4.
Predicted reactivity: Mouse (100%), Rat (100%), Zebrafish (89%), Xenopus laevis (93%), Bovine (99%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
NEK8 serine/threonine kinase or NIMA-related kinase 8 is related to NIMA (never in mitosis, gene A) of Aspergillus nodulins and may play a role in cell cycle progression from G2 to M phase. NEK8 is part of a complex of ciliary proteins required for renal and cardiovascular development. NEK8 interacts with polycystin signal transduction pathways and may control the targeting of these ciliary proteins. Required for renal tubular integrity. May regulate local cytoskeletal structure in kidney tubule epithelial cells. May regulate ciliary biogenesis through targeting of proteins to the cilia.Defects in NEK8 are the cause of nephronophthisis type 9 (NPHP9) [MIM:613824]. NPHP9 is an autosomal recessive disorder resulting in end-stage renal disease. It is a progressive tubulo-interstitial kidney disorder histologically characterized by modifications of the tubules with thickening of the basement membrane, interstitial fibrosis and, in the advanced stages, medullary cysts.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: NEK 8; nephrocystin 9; Never in mitosis A-related kinase 8; NIMA (never in mitosis gene a)- related kinase 8; NIMA-family kinase NEK8; NIMA-related kinase 12a; NIMA-related kinase 8; Nima-related protein kinase 12a; NimA-related protein kinase 8; Serine/threonine-protein kinase Nek8; serine/thrionine-protein kinase NEK8
Gene Aliases: JCK; NEK12A; NEK8; NPHP9; RHPD2
UniProt ID: (Human) Q86SG6
Entrez Gene ID: (Human) 284086
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support