Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MSVLGEYERH CDSINSDFGS ESGGCGDSSP GPSASQGPRA GGGAAEQEEL HYIPIRVLGR GAFGEATLYR RTEDDSLVVW KEVDLTRLSE KERRDALNEI VILALLQHDN IIAYYNHFMD NTTLLIELEY CNGGNLYDKI LRQKDKLFEE EMVVWYLFQI VSAVSCIHKA GILHRDIKTL NIFLTKANLI KLGDYGLAKK LNSEYSMAET LVGTPYYMSP ELCQGVKYNF KSDIWAVGCV IFELLTLKRT FDATNPLNLC VKIVQGIRAM EVDSSQYSLE LIQMVHSCLD QDPEQRPTAD ELLDRPLLRK RRREMEEKVT LLNAPTKRPR SSTVTEAPIA VVTSRTSEVY VWGGGKSTPQ KLDVIKSGCS ARQVCAGNTH FAVVTVEKEL YTWVNMQGGT KLHGQLGHGD KASYRQPKHV EKLQGKAIRQ VSCGDDFTVC VTDEGQLYAF GSDYYGCMGV DKVAGPEVLE PMQLNFFLSN PVEQVSCGDN HVVVLTRNKE VYSWGCGEYG RLGLDSEEDY YTPQKVDVPK ALIIVAVQCG CDGTFLLTQS GKVLACGLNE FNKLGLNQCM SGIINHEAYH EVPYTTSFTL AKQLSFYKIR TIAPGKTHTA AIDERGRLLT FGCNKCGQLG VGNYKKRLGI NLLGGPLGGK QVIRVSCGDE FTIAATDDNH IFAWGNGGNG RLAMTPTERP HGSDICTSWP RPIFGSLHHV PDLSCRGWHT ILIVEKVLNS KTIRSNSSGL SIGTVFQSSS PGGGGGGGGG EEEDSQQESE TPDPSGGFRG TMEADRGMEG LISPTEAMGN SNGASSSCPG WLRKELENAE FIPMPDSPSP LSAAFSESEK DTLPYEELQG LKVASEAPLE HKPQVEASSP RLNPAVTCAG KGTPLTPPAC ACSSLQVEVE RLQGLVLKCL AEQQKLQQEN LQIFTQLQKL NKKLEGGQQV GMHSKGTQTA KEEMEMDPKP DLDSDSWCLL GTDSCRPSL
NEK9, a NEK type protein kinase, regulates chromosome alignment and segregation in mitosis. The protein has a N-terminal NIMA-like catalytic domain, a central domain with homology to the guanine nucleotide exchange factor for the GTPase Ran (RCC1), and a C-terminal coiled-coil domain. It is phosphorylated by active p34(Cdc2) and is capable of autophosphorylation and oligomerization. The activated protein binds Nek6 and RCC1, and has been shown to associate with the human homolog of the Drosophila protein Bicaudal D (Bicd2) in vivo and to phosphorylate this coiled-coil protein in vitro. NEK9 expression has been documented in normal human brain. ESTs have been isolated from several tissue libraries, including normal bone, lymph and thymus and cancerous cervix, eye, lymph and placenta.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZp434D0935; MGC138306; MGC16714; NEK 9; Nek8; Nercc 1 kinase; Nercc1 kinase; Never in mitosis A-related kinase 9; NIMA (never in mitosis gene a)- related kinase 9; NimA-related kinase 8; NimA-related protein kinase 9; Serine/threonine-protein kinase Nek9
Gene Aliases: APUG; KIAA1995; LCCS10; NC; NEK8; NEK9; NERCC; NERCC1
UniProt ID: (Human) Q8TD19
Entrez Gene ID: (Human) 91754
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support