Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: MVGPAPRRRL RPLAALALVL ALAPGLPTAR AGQTPRPAER GPPVRLFTEE ELARYGGEEE DQPIYLAVKG VVFDVTSGKE FYGRGAPYNA LTGKDSTRGV AKMSLDPADL THDTTGLTAK ELEALDEVFT KVYKAKYPIV GYTARRILNE DGSPNLDFKP EDQPHFDIKD EF
NENF is a secreted protein that is expressed in neurons but not glial cells of the brain. This protein has neurotrophic activity in primary cultured mouse neurons but not mitogenic activity in primary cultured mouse astrocytes. NENF activated mitogen-activated protein (MAP) and phosphotidylinositol-3 kinase pathways and could be inhibited by pertussis toxin but not receptor tyrosine kinases, suggesting that NENF acts through a Gi/Go-protein-coupled receptor.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cell growth-inhibiting protein 47; Cell immortalization-related protein 2; Neudesin; Neuron-derived neurotrophic factor; Protein GIG47; SCIRP10-related protein; Secreted protein of unknown function; Spinal cord injury related protein 10; SPUF protein
Gene Aliases: CIR2; NENF; SCIRP10; SPUF
UniProt ID: (Human) Q9UMX5
Entrez Gene ID: (Human) 29937
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support