Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Aliquoting is unnecessary for -20°C storage.
This gene belongs to a family of glycohydrolytic enzymes which remove sialic acid residues from glycoproteins and glycolipids. Expression studies in COS7 cells confirmed that this gene encodes a functional sialidase. Its cytosolic localization was demonstrated by cell fractionation experiments.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: brain sialidase; cystolic sialidase; Cytosolic sialidase; Mouse skeletal muscle sialidase; MSS; MTS; Murine thymic sialidase; N-acetyl-alpha-neuraminidase 2; neuraminidase 2; neuraminidase 2 (cytosolic sialidase); sialidase 2 (cytosolic sialidase); Sialidase-2; skeletal muscle sialidase
Gene Aliases: MBS; MSS; MTS; NEU2; SIAL2
UniProt ID: (Human) Q9Y3R4, (Mouse) Q9JMH3
Entrez Gene ID: (Human) 4759, (Mouse) 23956, (Rat) 29204
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support