Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide sequence: EEEGLFGCLF ECGTKQECEQ IAFRLFTHRE ILSHLQGDCT SPGRNPSQFK
Sequence homology: Horse: 100%; Human: 100%
The function of this protein remains unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ganglioside sialidase; Ganglioside sialidasedis; Membrane sialidase; N-acetyl-alpha-neuraminidase 3; neuraminidase 3 (membrane sialidase); neuraminidase 3, membrane sialidase; sialidase 3 (membrane sialidase); Sialidase-3
Gene Aliases: NEU3; SIAL3
UniProt ID: (Human) Q9UQ49
Entrez Gene ID: (Human) 10825
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support