Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 5
Antibody detects endogenous levels of total NEUROD4.
Probably acts as a transcriptional activator. Mediates neuronal differentiation. Required for the regulation of amacrine cell fate specification in the retina.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATH-3; atonal homolog 3; bHLHa4; Class A basic helix-loop-helix protein 4; Helix-loop-helix protein mATH-3; mATH3; NeuroD4; neurogenic differentiation 4; Neurogenic differentiation factor 4; Protein atonal homolog 3
Gene Aliases: AI846749; ATH-3; ATH3; ATOH3; BHLHA4; MATH-3; MATH3; NEUROD4
UniProt ID: (Human) B2RAC9, (Mouse) O09105
Entrez Gene ID: (Human) 58158, (Mouse) 11923, (Rat) 288821
Molecular Function: basic helix-loop-helix transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support