Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: CRPARLLGLV HDCKRRPSRA RAVSRGAKTA ETVQRIKKTR RLKANNRERN RMHNLNAALD ALREVLPTFP EDAKLTKIET LRFAHNYIWA LTETLRLADH CG
Neurogenin-2 is a member of the neurogenin subfamily of basic helix-loop-helix (bHLH) transcription factor genes that play an important role in neurogenesis from migratory neural crest cells. In the course of mouse neurogenesis, Ngn2 and Ngn1 are expressed in distinct progenitor populations residing in the central and peripheral nervous systems. Mutation analyses demonstrate that Ngn1 is required for the determination of neuronal precursors for proximal cranial sensory ganglia and that Ngn2 is necessary for the determination of precursors for epibranchial placode-derived sensory neurons.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: bHLHa8; Class A basic helix-loop-helix protein 8; Neurogenin-2; neurogenin2; NGN-2; Protein atonal homolog 4
Gene Aliases: ATOH4; BHLHA8; Math4A; NEUROG2; ngn-2; NGN2
UniProt ID: (Human) Q9H2A3
Entrez Gene ID: (Human) 63973
Molecular Function:
basic helix-loop-helix transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support