Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 5
Synthetic peptide sequence is EYQDLLNVKMALDIEIAAYRKLLEGEE
Neurofilaments are type IV intermediate filament heteropolymers composed of light, medium, and heavy chains. Neurofilaments comprise the axoskeleton and functionally maintain neuronal caliber. They may also play a role in intracellular transport to axons and dendrites. This gene encodes the medium neurofilament protein. This protein is commonly used as a biomarker of neuronal damage. Alternative splicing results in multiple transcript variants encoding distinct isoforms. Diseases associated with NEFM include Pineal Parenchymal Tumor Of Intermediate Differentiation and Wallerian Degeneration. Among its related pathways are Amyotrophic lateral sclerosis (ALS) and Neural Stem Cell Differentiation Pathways and Lineage-specific Markers. Gene Ontology (GO) annotations related to this gene include structural molecule activity and structural constituent of cytoskeleton.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 160 kDa neurofilament protein; 200 kDa neurofilament protein; 68 kDa neurofilament protein; light molecular weight neurofilament protein; neurofilament; Neurofilament 3; neurofilament heavy; Neurofilament heavy polypeptide; neurofilament light; Neurofilament light polypeptide; Neurofilament medium polypeptide; neurofilament protein, light chain; neurofilament subunit NF-L; Neurofilament triplet H protein; Neurofilament triplet L protein; Neurofilament triplet M protein; neurofilament, heavy polypeptide 200kDa; neurofilament, light polypeptide 68kDa; neurofilament, medium polypeptide 150kDa; neurofilament-3; neurofilament-3 (150 kD medium); neurofilament-l; NF-H; NF-L; NF-M; NHC; NLC; protein phosphatase 1, regulatory subunit 110
Gene Aliases: CMT1F; CMT2CC; CMT2E; KIAA0845; NEF3; NEFH; NEFL; NEFM; NF-L; NF-M; NF68; NFH; NFL; NFM; PPP1R110
UniProt ID: (Human) P12036, (Human) P07196, (Human) P07197
Entrez Gene ID: (Human) 4744, (Human) 4747, (Human) 4741
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support