Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody recognize NEFH only.
Immunogen sequence: MMSFGGADA LLGAPFAPLH GGGSLHYALA RKGGAGGTRS AAGSSSGFHS WTRTSVSSVS ASPSRFRGAG AASSTDSLDT LSNGPEGCMV AVATSRSEKE QLQALNDRFA GYIDKVRQLE AHNRSLEGEA AALRQQQAGR SAMGELYERE VREMRGAVLR LGAARGQLRL EQEHLLEDIA HVRQRLDDEA RQREEAEAAA RALARFAQEA EAARV (1-214 aa encoded by BC014185)
Involved in the maintenance of neuronal caliber, neurofilaments are the intermediate filament proteins found specifically in neurons, and are composed predominantly of three major proteins called NF-L, NF-M and NF-H. Like most other intermediate filament proteins (IFPs), the expression of the different neuronal IFPs is both tissue-specific and developmentally regulated. NF-H has an important role in mature axons and is involved in amyotrophic lateral sclerosis (ALS).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 200 kDa neurofilament protein; NEF3; neurofilament; neurofilament 200kDa; Neurofilament heavy polypeptide; Neurofilament triplet H protein; neurofilament, heavy polypeptide 200kDa; NF 200 kD; NF-H; NHC
Gene Aliases: CMT2CC; KIAA0845; mKIAA0845; NEFH; NF-H; NF200; NFH
UniProt ID: (Human) P12036, (Mouse) P19246
Entrez Gene ID: (Human) 4744, (Pig) 100156492, (Rat) 24587, (Mouse) 380684
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support