Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSSFSYEPYY STSYKRRYVE TPRVHISSVR SGYSTARSAY SSYSAPVSSS LSVRRSYSSS SGSLMPSLEN LDLSQVAAIS NDLKSIRTQE KAQLQDLNDR FASFIERVHE LEQQNKVLEA ELLVLRQKHS EPSRFRALYE QEIRDLRLAA EDATNEKQAL QGEREGLEET LRNLQARYEE EVLSREDAEG RLMEARKGAD EAALARAELE KRIDSLMDEI SFLKKVHEEE IAELQAQIQY AQISVEMDVT KPDLSAALKD IRAQYEKLAA KNMQNAEEWF KSRFTVLTES AAKNTDAVRA AKDEVSESRR LLKAKTLEIE ACRGMNEALE KQLQELEDKQ NADISAMQDT IN
Involved in the maintenance of neuronal caliber, neurofilaments are the intermediate filament proteins found specifically in neurons, and are composed predominantly of three major proteins called NF-L, NF-M and NF-H. Like most other intermediate filament proteins (IFPs), the expression of the different neuronal IFPs is both tissue-specific and developmentally regulated. NF-L is the light or low molecular weight microfilament subunit and runs on SDS-PAGE gels at approximately 70 kDa. Neurofilament are the 10nm or intermediate filament proteins found specifically in neurons, and are composed predominantly of three major proteins called NF-L, NF-M and NF-H. NF-H is the heavy or high molecular weight microfilament subunit and runs on SDS-PAGE gels in the range 180-220 kDa, with some variation in different species.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 68 kDa neurofilament protein; FLJ53642; light molecular weight neurofilament protein; NEF3; neurofilament; neurofilament 3; Neurofilament light polypeptide; neurofilament protein L; neurofilament protein, light chain; neurofilament subunit NF-L; Neurofilament triplet L protein; neurofilament, light polypeptide; neurofilament, light polypeptide 68kDa; neurofilament-l; neurofilament-light; NF-L; NFM; NLC; OTTHUMP00000196614; protein phosphatase 1, regulatory subunit 110
Gene Aliases: AI847934; CMT1F; CMT2E; NEFL; NF-L; NF68; NFL; PPP1R110
UniProt ID: (Human) P07196, (Mouse) P08551, (Rat) P19527
Entrez Gene ID: (Human) 4747, (Pig) 100521224, (Mouse) 18039, (Rat) 83613
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support