FIGURE: 1 / 3
Immunogen sequence: VNDDRHVMAK HSSVYPTQEE LEAVQNMVSH TERALKAVSD WIDEQEKGSS EQAESDNMDV PPEDDSKEGA GEQKTEHMTR TLRGVMRVGL VAKGLLLKGD LDLELVLLCK EKPTTALLDK VADNLAIQLA AVTEDKYEIL QSVDDAAIVI KNTKEPPLSL TIHLTSPVVR EEMEKVLAGE TLSVNDPPDV LDRQKCLAAL ASLRHAKWFQ ARANGLKSCV IVIRVLRDLC TRVPTWGPLR GWPLELLCEK SIGTANRPMG AGEALRRVLE CLASGIVMPD GSGIYDPCEK EATDAIGHLD RQQREDITQS AQHALRLAAF GQLHKVLGMD PLPSKM
UBA6 (UBE1L2, MOP-4)is a novel E1 enzyme specific for ubiquitin and belongs to the ubiquitin-activating E1 family. It is a part of the cascade to attach ubiquitin (Ub) covalently to substrate proteins. UBA6 activates ubiquitin by first adenylating its carboxy-terminal glycine residue with ATP and then linking this residue to the side chain of a cysteine residue in E1, yielding an ubiquitin-E1 thioester and free AMP. Its isoforms influence spermatogenesis and male fertility. UBA6 is highly expressed in human monocytes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Double-stranded RNA-binding protein 76; double-stranded RNA-binding protein, 76 kD; DRBP76; dsRNA binding protein NFAR-2/MPP4; interleukin enhancer binding factor 3, 90kD; interleukin enhancer binding factor 3, 90kDa; Interleukin enhancer-binding factor 3; M phase phosphoprotein 4; M-phase phosphoprotein 4; MPP4; NF-AT-90; NFAR; nuclear factor 90; Nuclear factor associated with dsRNA; Nuclear factor of activated T-cells 90 kDa; nuclear factor of activated T-cells, 90 kD; TCP80; Translational control protein 80
Gene Aliases: CBTF; DRBF; DRBP76; ILF3; MBII-26; MMP4; MPHOSPH4; MPP4; NF-AT-90; NF110; NF110b; NF90; NF90a; NF90b; NFAR; NFAR-1; NFAR2; TCP110; TCP80
UniProt ID: (Human) Q9NQ95, (Mouse) Q9Z1X4, (Rat) Q9JIL3
Entrez Gene ID: (Human) 3609, (Mouse) 16201, (Rat) 84472
Molecular Function:
RNA metabolism protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support