Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The recommended shelf life for this product is 1 year from date of receipt.
Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
This protein is an adapter or scaffold protein, which participates in the assembly of numerous protein complexes and is involved in several cellular processes such as cell fate determination, cytoskeletal organization, repression of gene transcription, mitosis, cell-cell adhesion, cell differentiation, proliferation and migration.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: NF-AT3; NF-ATc4; NFATc4; Nuclear factor of activated T-cells, cytoplasmic 4; nuclear factor of activated T-cells, cytoplasmic, calcineurin-dependent 4; T-cell transcription factor NFAT3; transcription complex subunit NF-ATc4
Gene Aliases: 3110041H08Rik; AW107667; AW546455; NF-AT3; NF-ATC4; NFAT3; NFATC4
UniProt ID: (Human) Q14934, (Mouse) Q8K120
Entrez Gene ID: (Human) 4776, (Mouse) 73181
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support