Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Positive Samples: K562, mouse intestine; Cellular Location: Cytoplasm, Nucleus
Immunogen sequence: MNAPERQPQP DGGDAPGHEP GGSPQDELDF SILFDYEYLN PNEEEPNAHK VASPPSGPAY PDDVLDYGLK PYSPLASLSG EPPGRFGEPD RVGPQKFLSA AKPAGASGLS PRIEITPSHE LIQAVGPLRM RDAGLLVEQP PLAGVAASPR FTLPVPGFEG YREPLCLSPA SSGSSASFIS DTFSPYTSPC VSPNNGGPDD LCPQFQNIPA HYSPRTSPIM SPRTSLAEDS CLGRHSPVPR PASRSSSPGA KRRHSCAEAL VALPPGASPQ RSRSPSPQPS SHVAPQDHGS PAGYPPVAGS
The nuclear factor of activated T-cells (NFAT) transcription complex is required for the expression of a group of proteins that collectively regulate the immune response. Four NFAT proteins, encoded on separate genes and expressed as several splice variants, have been described: NFAT1 (also known as NFATp or NFATc2), NFAT2 (NFATc or NFATc1), NFAT3, and NFAT4 (NFATx or NFATc3). These proteins show a low level of sequence similarity with the Dorsal/Rel/NFkB family of transcription factors. Another NFAT-related protein termed NFAT5 differs from isoforms 1-4 in that it lacks many of the Fos/Jun contact sites observed in its predecessors and its subcellular localization is not calcineurin-dependent.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: IB-IXL; KIAA0611; NF-ATc2; NF-ATp; NFAT pre-existing s; NFAT pre-existing subunit; NFAT transcription complex, preexisting component; Nuclear factor of activated T-cells, cytoplasmic 2; nuclear factor of activated T-cells, cytoplasmic, calcineurin-dependent 2; nuclear factor of activated T-cells, preexisting component; OTTHUMP00000031291; preexisting nuclear factor of activated T-cells 2; T cell transcription factor NFAT1; T-cell transcription factor NFAT1
Gene Aliases: AI607462; NF-ATc2; NF-ATp; NFAT1; NFAT1-D; NFATC2; NFATP
UniProt ID: (Human) Q13469, (Mouse) Q60591
Entrez Gene ID: (Human) 4773, (Rat) 311658, (Mouse) 18019
Molecular Function: Rel homology transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support