Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: MSEPLQSVVD HMATHLGVSP SRILLLFGET ELSPTATPRT LKLGVADIID CVVLTSSPEA TETSQQLQLR VQGKEKHQTL EVSLSRDSPL KTLMSHYEEA MGLSGRKLSF FFDGTKLSGR ELPADLGMES GDLIEVWG
In T-helper 2 (Th2) cells, regulates the magnitude of NFAT-driven transcription of a specific subset of cytokine genes, including IL3, IL4, IL5 and IL13, but not IL2. Recruits PRMT1 to the IL4 promoter; this leads to enhancement of histone H4 'Arg-3'-methylation and facilitates subsequent histone acetylation at the IL4 locus, thus promotes robust cytokine expression. Down-regulates formation of poly-SUMO chains by UBE2I/UBC9.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 45 kDa NF-AT-interacting protein; 45 kDa NFAT-interacting protein; NFATC2-interacting protein; Nuclear factor of activated T-cells, cytoplasmic 2-interacting protein; nuclear factor of activated T-cells, cytoplasmic, calcineurin-dependent 2 interacting protein
Gene Aliases: ESC2; NFATC2IP; NIP45; RAD60
UniProt ID: (Human) Q8NCF5
Entrez Gene ID: (Human) 84901
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support