Product References
miR-339-3p promotes AT1-AA-induced vascular inflammation by upregulating NFATc3 protein expression in vascular smooth muscle cells.
Acta biochimica et biophysica Sinica
Li Y,Long Y,Zhi X,Hao H,Wang X,Liu H,Wang L
Sat Feb 25 00:00:00 EST 2023
Blocking Store-Operated Ca2+ Entry to Protect HL-1 Cardiomyocytes from Epirubicin-Induced Cardiotoxicity.
Cells
Liu X,Chang Y,Choi S,Cai C,Zhang X,Pan Z
Fri Feb 24 00:00:00 EST 2023
Cullin-associated and neddylation-dissociated 1 protein (CAND1) governs cardiac hypertrophy and heart failure partially through regulating calcineurin degradation.
Pharmacological research
Li X,Zhang Y,Zhao Y,Zhou Y,Han Q,Yang Y,Zhang L,Shi L,Jin X,Zhang R,Gao H,Xue G,Li D,Zhang ZR,Lu Y,Yang B,Pan Z
Mon Aug 01 00:00:00 EDT 2022
Trans-cinnamaldehyde suppresses microtubule detyrosination and alleviates cardiac hypertrophy.
European journal of pharmacology
Tian J,Shan XL,Wang SN,Chen HH,Zhao P,Qian DD,Xu M,Guo W,Zhang C,Lu R
Wed Jan 05 00:00:00 EST 2022
TARBP2-stablized SNHG7 regulates blood-brain barrier permeability by acting as a competing endogenous RNA to miR-17-5p/NFATC3 in Aβ-microenvironment.
Cell death & disease
Ning H,Zhang L,Zhu B,Zhou X,Zhang T,Ma T
Fri May 13 00:00:00 EDT 2022
A-kinase anchoring protein 5-ancored calcineurin regulates the remodeling of H9c2 cardiomyocytes exposed to hypoxia and reoxygenation.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Zhu F,Yuan C,Zhang X,Wang Z,Wang Q,Wang H
Tue Nov 01 00:00:00 EDT 2022
Substitution of SERCA2 Cys674 aggravates cardiac fibrosis by promoting the transformation of cardiac fibroblasts to cardiac myofibroblasts.
Biochemical pharmacology
Huang X,Lin X,Wang L,Xie Y,Que Y,Li S,Hu P,Tong X
Thu Sep 01 00:00:00 EDT 2022
Trim39 regulates neuronal apoptosis by acting as a SUMO-targeted E3 ubiquitin-ligase for the transcription factor NFATc3.
Cell death and differentiation
Basu-Shrivastava M,Mojsa B,Mora S,Robbins I,Bossis G,Lassot I,Desagher S
Tue Nov 01 00:00:00 EDT 2022
Trim39 regulates neuronal apoptosis by acting as a SUMO-targeted E3 ubiquitin-ligase for the transcription factor NFATc3.
Cell death and differentiation
Basu-Shrivastava M,Mojsa B,Mora S,Robbins I,Bossis G,Lassot I,Desagher S
Tue Nov 01 00:00:00 EDT 2022
Pou3f1 mediates the effect of Nfatc3 on ulcerative colitis-associated colorectal cancer by regulating inflammation.
Cellular & molecular biology letters
Lin Y,Wang D,Zhao H,Li D,Li X,Lin L
Mon Sep 05 00:00:00 EDT 2022
Pou3f1 mediates the effect of Nfatc3 on ulcerative colitis-associated colorectal cancer by regulating inflammation.
Cellular & molecular biology letters
Lin Y,Wang D,Zhao H,Li D,Li X,Lin L
Mon Sep 05 00:00:00 EDT 2022
Metformin inhibition of mitochondrial ATP and DNA synthesis abrogates NLRP3 inflammasome activation and pulmonary inflammation.
Immunity
Xian H,Liu Y,Rundberg Nilsson A,Gatchalian R,Crother TR,Tourtellotte WG,Zhang Y,Aleman-Muench GR,Lewis G,Chen W,Kang S,Luevanos M,Trudler D,Lipton SA,Soroosh P,Teijaro J,de la Torre JC,Arditi M,Karin M,Sanchez-Lopez E
Tue Jul 13 00:00:00 EDT 2021
Regulator of calcineurin 1 gene isoform 4 in pancreatic ductal adenocarcinoma regulates the progression of tumor cells.
Oncogene
Lao M,Zhang X,Ma T,Xu J,Yang H,Duan Y,Ying H,Zhang X,Guo C,Qiu J,Bai X,Liang T
Thu Apr 01 00:00:00 EDT 2021
TRPC3 promotes tumorigenesis of gastric cancer via the CNB2/GSK3β/NFATc2 signaling pathway.
Cancer letters
Lin DC,Zheng SY,Zhang ZG,Luo JH,Zhu ZL,Li L,Chen LS,Lin X,Sham JSK,Lin MJ,Zhou RX
Thu Oct 28 00:00:00 EDT 2021
The role of nuclear receptor 4A1 (NR4A1) in drug-induced gingival overgrowth.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Hatano S,Matsuda S,Okanobu A,Furutama D,Memida T,Kajiya M,Ouhara K,Fujita T,Mizuno N,Kurihara H
Thu Jul 01 00:00:00 EDT 2021
The role of nuclear receptor 4A1 (NR4A1) in drug-induced gingival overgrowth.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
Hatano S,Matsuda S,Okanobu A,Furutama D,Memida T,Kajiya M,Ouhara K,Fujita T,Mizuno N,Kurihara H
Thu Jul 01 00:00:00 EDT 2021
Transmembrane Protein Ttyh1 Maintains the Quiescence of Neural Stem Cells Through Ca2+/NFATc3 Signaling.
Frontiers in cell and developmental biology
Cao Y,Wu HN,Cao XL,Yue KY,Han WJ,Cao ZP,Zhang YF,Gao XY,Luo C,Jiang XF,Han H,Zheng MH
Tue Dec 07 00:00:00 EST 2021
Protective Role of Tangshen Formula on the Progression of Renal Damage in db/db Mice by TRPC6/Talin1 Pathway in Podocytes.
Journal of diabetes research
Wang Q,Tian X,Zhou W,Wang Y,Zhao H,Li J,Zhou X,Zhang H,Zhao T,Li P
Mon Aug 23 00:00:00 EDT 2021
Methionine Enkephalin Suppresses Osteocyte Apoptosis Induced by Compressive Force through Regulation of Nuclear Translocation of NFATc1.
JBMR plus
Sogi C,Takeshita N,Jiang W,Kim S,Maeda T,Yoshida M,Oyanagi T,Ito A,Kimura S,Seki D,Takano I,Sakai Y,Fujiwara I,Kure S,Takano-Yamamoto T
Wed Jul 01 00:00:00 EDT 2020
The calcium pump PMCA4 prevents epithelial-mesenchymal transition by inhibiting NFATc1-ZEB1 pathway in gastric cancer.
Biochimica et biophysica acta. Molecular cell research
Wang T,Li N,Jin L,Qi X,Zhang C,Hua D
Tue Dec 01 00:00:00 EST 2020
NFATc3 regulation of collagen V expression contributes to cellular immunity to collagen type V and hypoxic pulmonary hypertension.
American journal of physiology. Lung cellular and molecular physiology
Sheak JR,Jones DT,Lantz BJ,Maston LD,Vigil D,Resta TC,Resta MM,Howard TA,Kanagy NL,Guo Y,Jankowska-Gan E,Sullivan JA,Braun RK,Burlingham WJ,Gonzalez Bosc LV
Tue Dec 01 00:00:00 EST 2020
NFATc3 regulation of collagen V expression contributes to cellular immunity to collagen type V and hypoxic pulmonary hypertension.
American journal of physiology. Lung cellular and molecular physiology
Sheak JR,Jones DT,Lantz BJ,Maston LD,Vigil D,Resta TC,Resta MM,Howard TA,Kanagy NL,Guo Y,Jankowska-Gan E,Sullivan JA,Braun RK,Burlingham WJ,Gonzalez Bosc LV
Tue Dec 01 00:00:00 EST 2020
Tsantan Sumtang Restored Right Ventricular Function in Chronic Hypoxia-Induced Pulmonary Hypertension Rats.
Frontiers in pharmacology
Yang Z,Sun H,Su S,Nan X,Li K,Jin X,Jin G,Li Z,Lu D
Sat Feb 06 00:00:00 EST 2021
Stachydrine hydrochloride suppresses phenylephrine-induced pathological cardiac hypertrophy by inhibiting the calcineurin/nuclear factor of activated T-cell signalling pathway.
European journal of pharmacology
Zheng J,Tian J,Wang S,Hu P,Wu Q,Shan X,Zhao P,Zhang C,Guo W,Xu M,Chen H,Lu R
Tue Sep 15 00:00:00 EDT 2020
Blocking NFATc3 ameliorates azoxymethane/dextran sulfate sodium induced colitis-associated colorectal cancer in mice via the inhibition of inflammatory responses and epithelial-mesenchymal transition.
Cellular signalling
Lin Y,Koumba MH,Qu S,Wang D,Lin L
Thu Oct 01 00:00:00 EDT 2020
TRPV2-induced Ca2+-calcineurin-NFAT signaling regulates differentiation of osteoclast in multiple myeloma.
Cell communication and signaling : CCS
Bai H,Zhu H,Yan Q,Shen X,Lu X,Wang J,Li J,Chen L
Tue Oct 16 00:00:00 EDT 2018
TRPV2-induced Ca2+-calcineurin-NFAT signaling regulates differentiation of osteoclast in multiple myeloma.
Cell communication and signaling : CCS
Bai H,Zhu H,Yan Q,Shen X,Lu X,Wang J,Li J,Chen L
Tue Oct 16 00:00:00 EDT 2018
TRPV2-induced Ca2+-calcineurin-NFAT signaling regulates differentiation of osteoclast in multiple myeloma.
Cell communication and signaling : CCS
Bai H,Zhu H,Yan Q,Shen X,Lu X,Wang J,Li J,Chen L
Tue Oct 16 00:00:00 EDT 2018
TRPV2-induced Ca2+-calcineurin-NFAT signaling regulates differentiation of osteoclast in multiple myeloma.
Cell communication and signaling : CCS
Bai H,Zhu H,Yan Q,Shen X,Lu X,Wang J,Li J,Chen L
Tue Oct 16 00:00:00 EDT 2018
Calcineurin/NFAT Signaling Modulates Pulmonary Artery Smooth Muscle Cell Proliferation, Migration and Apoptosis in Monocrotaline-Induced Pulmonary Arterial Hypertension Rats.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology
He RL,Wu ZJ,Liu XR,Gui LX,Wang RX,Lin MJ
Tue Sep 18 00:00:00 EDT 2018
Calcineurin/NFAT Signaling Modulates Pulmonary Artery Smooth Muscle Cell Proliferation, Migration and Apoptosis in Monocrotaline-Induced Pulmonary Arterial Hypertension Rats.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology
He RL,Wu ZJ,Liu XR,Gui LX,Wang RX,Lin MJ
Tue Sep 18 00:00:00 EDT 2018
Regulator of Calcineurin 1 Gene Isoform 4, Down-regulated in Hepatocellular Carcinoma, Prevents Proliferation, Migration, and Invasive Activity of Cancer Cells and Metastasis of Orthotopic Tumors by Inhibiting Nuclear Translocation of NFAT1.
Gastroenterology
Jin H,Wang C,Jin G,Ruan H,Gu D,Wei L,Wang H,Wang N,Arunachalam E,Zhang Y,Deng X,Yang C,Xiong Y,Feng H,Yao M,Fang J,Gu J,Cong W,Qin W
Fri Sep 01 00:00:00 EDT 2017
Regulator of Calcineurin 1 Gene Isoform 4, Down-regulated in Hepatocellular Carcinoma, Prevents Proliferation, Migration, and Invasive Activity of Cancer Cells and Metastasis of Orthotopic Tumors by Inhibiting Nuclear Translocation of NFAT1.
Gastroenterology
Jin H,Wang C,Jin G,Ruan H,Gu D,Wei L,Wang H,Wang N,Arunachalam E,Zhang Y,Deng X,Yang C,Xiong Y,Feng H,Yao M,Fang J,Gu J,Cong W,Qin W
Fri Sep 01 00:00:00 EDT 2017
Nuclear factor of activated T-cell c3 inhibition of mammalian target of rapamycin signaling through induction of regulated in development and DNA damage response 1 in human intestinal cells.
Molecular biology of the cell
Zhou Y,Wang Q,Guo Z,Weiss HL,Evers BM
Wed Aug 01 00:00:00 EDT 2012
Nuclear factor of activated T-cell c3 inhibition of mammalian target of rapamycin signaling through induction of regulated in development and DNA damage response 1 in human intestinal cells.
Molecular biology of the cell
Zhou Y,Wang Q,Guo Z,Weiss HL,Evers BM
Wed Aug 01 00:00:00 EDT 2012