Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The recommended shelf life for this product is 1 year from date of receipt.
This gene encodes a protein that is part of a post-splicing multiprotein complex involved in both mRNA nuclear export and mRNA surveillance. mRNA surveillance detects exported mRNAs with truncated open reading frames and initiates nonsense-mediated mRNA decay (NMD). When translation ends upstream from the last exon-exon junction, this triggers NMD to degrade mRNAs containing premature stop codons. This protein is located only in the cytoplasm. When translation ends, it interacts with the protein that is a functional homolog of yeast Upf2p to trigger mRNA decapping. Use of multiple polyadenylation sites has been noted for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: NF-AT4; NF-ATc3; NFAC3; NFATx; NFC3; nuclear factor of activated T-cells c3 isoform IE-Xa; Nuclear factor of activated T-cells, cytoplasmic 3; nuclear factor of activated T-cells, cytoplasmic, calcineurin-dependent 3; T cell transcription factor NFAT4; T-cell transcription factor NFAT4
Gene Aliases: NFAT4; NFATC3; NFATX
UniProt ID: (Human) Q12968
Entrez Gene ID: (Human) 4775
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support