Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody is a rabbit polyclonal antibody raised against a region of human NFE2L1.
Immunogen sequence: MQAMEVNTS ASEILYSAPP GDPLSTNYSL APNTPINQNV SLHQASLGGC SQDFLLFSPE VESLPVASSS TLLPLAPSNS TSLNSTFGST NLTGLFFPPQ LNGTANDTAG PELPDPLGGL LDEAMLDEIS LMDLAIEEGF NPVQASQLEE EFDSDSGLSL DSSHSPSSLS SSEGSSSSSS SSSSSSSSAS SSASSSFSEE GAVGYSSDSE TLDLEEAEGA VGYQPEYSKF CRMSYQDPAQ LSCLPYLEHV GHNHTYNMAP SALDSADLPP PSALKKGSKE KQADFLDKQM SRDEHRARAM KIPFTNDKII NLPVEEFNEL LSKYQLSEAQ LSLIRDIRRR GKNKMAAQNC RKRKLDTILN LERDVEDLQR DKARLLREKV EFLRSLRQMK QKVQSLYQEV FGRLRDENGR PYSPSQYALQ YAGDGSVLLI PRTMADQQAR RQERKPKDRR K (293-772 aa encoded by BC010623)
This gene encodes a protein that is involved in globin gene expression in erythrocytes. Confusion has occurred in bibliographic databases due to the shared symbol of NRF1 for this gene, NFE2L1, and for 'nuclear respiratory factor 1' which has an official symbol of NRF1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Endoplasmic reticulum membrane sensor NFE2L1; LCR-F1; Locus control region-factor 1; NF-E2-related factor 1; NFE2-related factor 1; nuclear factor (erythroid-derived 2)-like 1; nuclear factor erythroid 2-like 1; Nuclear factor erythroid 2-related factor 1; nuclear factor, erythroid 2-like 1; Nuclear factor, erythroid derived 2, like 1; nuclear factor, erythroid derived 2,-like 1; Protein NRF1, p120 form; TCF-11; transc; Transcription factor 11; transcription factor 11 (basic leucine zipper type); transcription factor HBZ17; transcription factor LCR-F1
Gene Aliases: AA408798; AW212678; HBZ17; LCR-F1; Lcrf1; NFE2L1; NRF1; TCF-11; TCF11
UniProt ID: (Human) Q14494, (Mouse) Q61985
Entrez Gene ID: (Human) 4779, (Mouse) 18023, (Rat) 360610
Molecular Function:
basic leucine zipper transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support