Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MYSPLCLTQD EFHPFIEALL PHVRAFAYTW FNLQARKRKY FKKHEKRMSK EEERAVKDEL LSEKPEVKQK WASRLLAKLR KDIRPEYRED FVLTVTGKKP PCCVLSNPDQ KGKMRRIDCL RQADKVWRLD LVMVILFKGI PLESTDGERL VKSPQCSNPG LCVQPHHIGV SVKELDLYLA YFVHAADSSQ SESPSQPSDA DIKDQPENGH LGFQDSFVTS GVFSVTELVR VSQTPIAAGT GPNFSLSDLE SSSYYSMSPG AMRRSLPSTS STSSTKRLKS VEDEMDSPGE EPFYTGQGRS PGSGSQSSGW HEVEPGMPSP TTLKKSEKSG FSSPSPSQTS SLGTAFTQHH RPVITGPRAS PHATPSTLHF PTSPIIQQPG PYFSHPAIRY HPQETLKEFV QLVCPDAGQQ AGQVGFLNPN GSSQGKVHNP FLPTPMLPPP PPPPMARPVP LPVPDTKPPT TSTEGGAASP TSPILVPGIK VAASHHPPDR PPDPFSTL
NF-1, also designated CTF, consists of a family of CCAAT box binding proteins that stimulate DNA replication and activate transcription. Analysis of human NF-1 messenger RNA has revealed two forms of the NF-1 protein arising from an alternate splicing of a single NF-1 gene. NF-1 binds its consensus DNA element as a homodimer via an amino-terminal DNA binding domain, and activates transcription through a putatively novel, proline-rich, carboxy terminal transactivation domain. The NF-1 protein has been shown to recognize and bind the adenovirus type 2 promoter and activate transcription of herpes simplex virus thymidine kinase genes. The NF-1 consensus element has been found in the upstream promoter region of myriad eukaryotic genes, including that of Ha-Ras, alpha-globin, HSP 70, GRP 78, Histone H1, myelin basic protein and in the Xenopus laevis vitellogenin gene promoter.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CCAAT-box-binding transcription factor; CTF; NF-I/A; NF1-A; Nuclear factor 1 A-type; Nuclear factor I/A; TGGCA-binding protein
Gene Aliases: CTF; KIAA1439; NF-I/A; NF1-A; NFI-A; NFI-L; NFIA
UniProt ID: (Human) B4DRJ3
Entrez Gene ID: (Human) 4774
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support