Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSTEGGFGG TSSSDAQQSL QSFWPRVMEE IRNLTVKDFR VQELPLARIK KIMKLDEDVK MISAEAPVLF AKAAQIFITE LTLRAWIHTE DNKRRTLQRN DIAMAITKFD QFDFLIDIVP RDELKPPKRQ EEVRQSVTPA EPVQYYFTLA QQPTAVQVQG QQQGQQTTSS TTTIQPGQII IAQPQQGQTT PVTMQVGEGQ QVQIVQAQPQ GQAQQAQSGT GQTMQVMQQI ITNTGEIQQI PVQLNAGQLQ YI (1-251 aa encoded by BC005003)
The CCAAT-binding factor NF-Y is a heteromeric transcription factor that specifically binds to CCAAT sequences in many eukaryotic genes. NF-Y is made up of three subunits, NF-YA, NF-YB and NF-YC. All three components are necessary for DNA binding. In each NF-Y subunit, the segment needed for formation of the NF-Y-DNA complex is conserved from yeast to human. These conserved segments are homologous to the histone-fold motif of eukaryotic histones. The DNA binding domains of the NF-YB and NF-YC subunits have been suggested to interact through a protein-protein histone-fold "handshake" motif in a manner analogous to the histone proteins, H2B and H2A, respectively.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CAAT box DNA-binding protein subunit C; CAAT-box DNA-binding protein subunit C; CBF-C; CCAAT binding factor of CBF-C/NFY-C; CCAAT binding factor subunit C; CCAAT transcription binding factor subunit gamma; CCAAT-binding transcription factor subunit C; DKFZp667G242; FLJ45775; histone H1 transcription factor large subunit 2A; NF-YC; Nuclear transcription factor Y subunit C; Nuclear transcription factor Y subunit gamma; nuclear transcription factor Y, gamma; transactivator HSM-1; Transactivator HSM-1/2; transcription factor NF-Y, C subunit
Gene Aliases: CBF-C; CBFC; H1TF2A; HAP5; HSM; NF-YC; NFYC
UniProt ID: (Human) Q9UML0, (Mouse) P70353, (Rat) Q62725
Entrez Gene ID: (Human) 4802, (Mouse) 18046, (Rat) 25337
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support