Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 20
Reconstitute in 100 µL of sterile water. Centrifuge to remove any insoluble material.
Specificity of this antibody: NG2.
A human melanoma-associated chondroitin sulfate proteoglycan plays a role in stabilizing cell-substratum interactions during early events of melanoma cell spreading on endothelial basement membranes. CSPG4 represents an integral membrane chondroitin sulfate proteoglycan expressed by human malignant melanoma cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AN2 proteoglycan; AN2/NG2 proteoglycan; Chondroitin sulfate proteoglycan 4; Chondroitin sulfate proteoglycan NG2; HMW MAA; MEL CSPG; nerve/glial antigen 2; Proteoglycan AN2
Gene Aliases: 4732461B14Rik; AN2; Cspg4; Kiaa4232; NG2
UniProt ID: (Mouse) Q8VHY0
Entrez Gene ID: (Mouse) 121021
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support