Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MERPEPELI RQSWRAVSRS PLEHGTVLFA RLFALEPDLL PLFQYNCRQF SSPEDCLSSP EFLDHIRKVM LVIDAAVTNV EDLSSLEEYL ASLGRKHRAV GVKLSSFSTV GESLLYMLEK CLGPAFTPAT RAAWSQLYGA VVQAMSRGWD GE (1-151 aa encoded by BC032509)
Globins are a superfamily of gas-binding heme proteins that are present in bacteria, protists, fungi, plants and animals. Globins play evolutionarily divergent roles which include binding, transport, scavenging, detoxification, and sensing of oxygen, nitric oxide and carbon monoxide. Neuroglobin (Ngb) is a hexacoordinate hemoglobin that is predominantly expressed in the vertebrate brain and may enhance oxygen supply to neural components. Neuroglobin displays a high affinity for oxygen and its presence in cerebral neurons suggests a role in neuronal responses to hypoxia or ischemia. For example, in vitro neuronal hypoxia causes an elevation in the levels of neuroglobin, which enhances neuronal cell survival. The human neuroglobin gene maps to chromosome 14q24 and encodes a 151 amino acid protein.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Neuroglobin; Nitrite reductase
Gene Aliases: NGB
UniProt ID: (Human) Q9NPG2
Entrez Gene ID: (Human) 58157
Molecular Function:
transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support